Assay Details
Target Gene Details
Entrez Gene ID: | 90025 |
Gene Name: | ubiquitin protein ligase E3D |
Gene Aliases: |
C6orf157, H10BH, UBE2CBP, YJR141W |
Location: |
Chr.6:82880834-83065841 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE3D | NM_001304437.1 | NP_001291366.1 | ||
| NM_198920.2 | NP_944602.1 | |||
| XM_011536238.2 | XP_011534540.1 | |||
| XM_011536242.1 | XP_011534544.1 | |||
| XM_017011458.1 | XP_016866947.1 | |||
| XM_017011460.1 | XP_016866949.1 | |||
| AB126062.1 | BAD01604.1 | |||
| AK298205.1 | ||||
| BC053645.1 | AAH53645.1 | |||
| BC057845.1 | AAH57845.1 | |||
| BC071763.1 | AAH71763.1 | |||
| BC101512.1 | AAI01513.1 | |||
| BC101538.1 | AAI01539.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1022371 | Chr.6:82581263 - 82913094 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs143163009,rs148150503] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map