Assay Details
Target Gene Details
Entrez Gene ID: | 51567 |
Gene Name: | tyrosyl-DNA phosphodiesterase 2 |
Gene Aliases: |
AD022, EAP2, EAPII, TTRAP, dJ30M3.3, hTDP2 |
Location: |
Chr.6:24649977-24666887 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TDP2 | NM_016614.2 | NP_057698.2 | ||
| AF201687.1 | AAG35600.1 | |||
| AF223469.1 | AAF64144.1 | |||
| AJ269473.1 | CAB92966.1 | |||
| AJ420495.1 | ||||
| AK002168.1 | BAA92119.1 | |||
| AK296623.1 | ||||
| AK298699.1 | ||||
| AK308836.1 | ||||
| BC017553.2 | AAH17553.1 | |||
| BC110375.1 | AAI10376.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv601164 | Chr.6:24624863 - 24661371 on Build GRCh38 | Gain |
|
| nsv601165 | Chr.6:24632414 - 24665779 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map