Assay Details
Target Gene Details
Entrez Gene ID: | 5269 |
Gene Name: | serpin family B member 6 |
Gene Aliases: |
CAP, DFNB91, MSTP057, PI-6, PI6, PTI, SPI3 |
Location: |
Chr.6:2948159-2972165 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINB6 | NM_001195291.2 | NP_001182220.2 | ||
| NM_001271822.1 | NP_001258751.1 | |||
| NM_001271823.1 | NP_001258752.1 | |||
| NM_001271824.1 | NP_001258753.1 | |||
| NM_001297699.1 | NP_001284628.1 | |||
| NM_001297700.1 | NP_001284629.1 | |||
| NM_004568.5 | NP_004559.4 | |||
| XM_011514672.1 | XP_011512974.1 | |||
| XM_011514674.2 | XP_011512976.1 | |||
| XM_011514676.2 | XP_011512978.1 | |||
| XM_017010940.1 | XP_016866429.1 | |||
| XM_017010941.1 | XP_016866430.1 | |||
| AK094983.1 | ||||
| AK098566.1 | ||||
| AK314578.1 | ||||
| BC098564.1 | AAH98564.1 | |||
| BG499326.1 | ||||
| BI259189.1 | ||||
| BI827451.1 | ||||
| BM789272.1 | ||||
| BP325314.1 | ||||
| BP370484.1 | ||||
| BQ681973.1 | ||||
| BX375950.2 | ||||
| BX538343.1 | CAD98106.1 | |||
| CD674931.1 | ||||
| CX782163.1 | ||||
| HY169891.1 | ||||
| Z22658.1 | CAA80373.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600832 | Chr.6:2945097 - 2970334 on Build GRCh38 | Loss |
|
| nsv830575 | Chr.6:2847376 - 3035682 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map