Assay Details
Target Gene Details
Entrez Gene ID: | 285753 |
Gene Name: | centrosomal protein 57 like 1 |
Gene Aliases: |
C6orf182, bA487F23.2, cep57R |
Location: |
Chr.6:109095130-109167696 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEP57L1 | NM_001083535.1 | NP_001077004.1 | ||
| NM_001271852.1 | NP_001258781.1 | |||
| NM_001271853.1 | NP_001258782.1 | |||
| XM_005266935.4 | XP_005266992.3 | |||
| XM_005266936.4 | XP_005266993.3 | |||
| XM_005266937.4 | XP_005266994.3 | |||
| XM_005266938.4 | XP_005266995.3 | |||
| XM_006715462.3 | XP_006715525.1 | |||
| XM_011535759.2 | XP_011534061.1 | |||
| XM_011535763.2 | XP_011534065.2 | |||
| XM_017010764.1 | XP_016866253.1 | |||
| XM_017010765.1 | XP_016866254.1 | |||
| XM_017010767.1 | XP_016866256.1 | |||
| XM_017010770.1 | XP_016866259.1 | |||
| XM_017010772.1 | XP_016866261.1 | |||
| AK092723.1 | ||||
| AK289910.1 | ||||
| BC064365.1 | AAH64365.1 | |||
| BM787514.1 | ||||
| BQ267680.1 |
More Information
Set Membership: |
|

Genomic Map