Assay Details
Target Gene Details
Entrez Gene ID: | 860 |
Gene Name: | runt related transcription factor 2 |
Gene Aliases: |
AML3, CBF-alpha-1, CBFA1, CCD, CCD1, CLCD, OSF-2, OSF2, PEA2aA, PEBP2aA |
Location: |
Chr.6:45328142-45664032 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RUNX2 | NM_001015051.3 | NP_001015051.3 | ||
| NM_001024630.3 | NP_001019801.3 | |||
| XM_011514960.2 | XP_011513262.1 | |||
| XM_011514961.2 | XP_011513263.1 | |||
| XM_011514962.2 | XP_011513264.1 | |||
| XM_011514963.2 | XP_011513265.1 | |||
| XM_011514964.2 | XP_011513266.1 | |||
| XM_011514965.2 | XP_011513267.1 | |||
| XM_017011391.1 | XP_016866880.1 | |||
| XM_017011393.1 | XP_016866882.1 | |||
| XM_017011394.1 | XP_016866883.1 | |||
| XM_017011395.1 | XP_016866884.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs75953810] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map