Assay Details
Target Gene Details
Entrez Gene ID: | 57505 |
Gene Name: | alanyl-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
AARSL, COXPD8, LKENP, MT-ALARS, MTALARS |
Location: |
Chr.6:44297850-44313358 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AARS2 | XM_011514764.2 | 22 | 2923 | XP_011513066.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv10641n54 | Chr.6:44224183 - 44314579 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map