Assay Details
Target Gene Details
Entrez Gene ID: | 6568 |
Gene Name: | solute carrier family 17 member 1 |
Gene Aliases: |
NAPI-1, NPT-1, NPT1 |
Location: |
Chr.6:25723744-25832108 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC17A1 | XM_017011201.1 | XP_016866690.1 |
Target Gene Details
Entrez Gene ID: | 10050 |
Gene Name: | solute carrier family 17 member 4 |
Gene Aliases: |
KAIA2138 |
Location: |
Chr.6:25754673-25781175 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC17A4 | NM_001286121.1 | NP_001273050.1 | ||
| NM_005495.2 | NP_005486.1 | |||
| XM_011514213.2 | XP_011512515.1 | |||
| XM_011514214.2 | XP_011512516.1 | |||
| XM_011514215.2 | XP_011512517.1 | |||
| XM_011514217.2 | XP_011512519.1 | |||
| XM_011514219.2 | XP_011512521.1 | |||
| XM_017010150.1 | XP_016865639.1 | |||
| XM_017010151.1 | XP_016865640.1 | |||
| AB020527.1 | BAA76663.1 | |||
| AK024903.1 | ||||
| AK293354.1 |
More Information
Set Membership: |
|

Genomic Map