Assay Details
Target Gene Details
Entrez Gene ID: | 11116 |
Gene Name: | FGFR1 oncogene partner |
Gene Aliases: |
FOP |
Location: |
Chr.6:166999317-167042418 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGFR1OP | NM_001278690.1 | NP_001265619.1 | ||
| NM_007045.3 | NP_008976.1 | |||
| NM_194429.2 | NP_919410.1 | |||
| AJ420472.1 | ||||
| AK289846.1 | ||||
| AK294950.1 | ||||
| AK312791.1 | ||||
| BC011902.2 | AAH11902.1 | |||
| DC418208.1 | ||||
| Y18046.1 | CAA77020.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv605260 | Chr.6:166570606 - 167041651 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map