Assay Details
Target Gene Details
Entrez Gene ID: | 114801 |
Gene Name: | transmembrane protein 200A |
Gene Aliases: |
KIAA1913, TTMA, TTMC |
Location: |
Chr.6:130366281-130443424 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM200A | NM_001258276.1 | NP_001245205.1 | ||
| NM_001258277.1 | NP_001245206.1 | |||
| XM_005266816.2 | XP_005266873.1 | |||
| XM_011535419.2 | XP_011533721.1 | |||
| AB067500.1 | ||||
| AK315824.1 | ||||
| BC044246.1 | AAH44246.1 | |||
| DA657363.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2732698 | Chr.6:130389370 - 130390105 on Build GRCh38 | Deletion |
|
| nsv604663 | Chr.6:129482340 - 130671010 on Build GRCh38 | Gain |
|

Genomic Map