Assay Details
Target Gene Details
Entrez Gene ID: | 51104 |
Gene Name: | abhydrolase domain containing 17B |
Gene Aliases: |
C9orf77, CGI-67, FAM108B1 |
Location: |
Chr.9:71859461-71911245 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD17B | NM_016014.3 | NP_057098.2 | ||
| XM_006717134.3 | XP_006717197.2 | |||
| XM_017014787.1 | XP_016870276.1 | |||
| XM_017014790.1 | XP_016870279.1 | |||
| AF151825.1 | AAD34062.1 | |||
| CR746318.1 |
More Information
Set Membership: |
|

Genomic Map