Assay Details
Target Gene Details
Entrez Gene ID: | 5069 |
Gene Name: | pappalysin 1 |
Gene Aliases: |
ASBABP2, DIPLA1, IGFBP-4ase, PAPA, PAPP-A, PAPPA1 |
Location: |
Chr.9:116153752-116402321 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PAPPA | NM_002581.4 | NP_002572.2 | ||
| XM_017014784.1 | XP_016870273.1 | |||
| AK308154.1 | ||||
| BC078657.1 | AAH78657.1 | |||
| CB960120.1 | ||||
| X68280.1 | CAA48341.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv825063 | Chr.9:116150580 - 116157180 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map