Assay Details
Target Gene Details
Entrez Gene ID: | 54914 |
Gene Name: | focadhesin |
Gene Aliases: |
KIAA1797 |
Location: |
Chr.9:20658309-20995955 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOCAD | NM_017794.4 | NP_060264.4 | ||
| XM_017014855.1 | XP_016870344.1 | |||
| AK056522.1 | BAB71203.1 | |||
| AY139834.1 | AAN17740.1 |
Target Gene Details
Entrez Gene ID: | 101929548 |
Gene Name: | FOCAD antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:20683302-20684750 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOCAD-AS1 | NR_121601.1 | |||
| BX115825.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv613747 | Chr.9:20668316 - 20793446 on Build GRCh38 | Loss |
|
| nsv1028928 | Chr.9:20659494 - 20793947 on Build GRCh38 | Loss |
|
| esv2764150 | Chr.9:20659506 - 22349184 on Build GRCh38 | Gain+Loss |
|
| nsv1032837 | Chr.9:20647170 - 20692439 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map