Assay Details
Target Gene Details
Entrez Gene ID: | 54926 |
Gene Name: | ubiquitin conjugating enzyme E2 R2 |
Gene Aliases: |
CDC34B, E2-CDC34B, UBC3B |
Location: |
Chr.9:33817164-33920403 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE2R2 | NM_017811.3 | NP_060281.2 | ||
| XM_005251496.2 | XP_005251553.1 | |||
| XM_011517949.1 | XP_011516251.1 | |||
| XM_017014860.1 | XP_016870349.1 | |||
| AJ240087.1 | CAC80336.1 | |||
| AK000426.1 | BAA91156.1 | |||
| AK225880.1 | ||||
| BC004862.2 | AAH04862.1 | |||
| BC047584.1 | AAH47584.1 | |||
| CR457233.1 | CAG33514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv520467 | Chr.9:33804815 - 34146647 on Build GRCh38 | Gain |
|
| nsv518977 | Chr.9:33675631 - 33845081 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map