Assay Details
Target Gene Details
Entrez Gene ID: | 5253 |
Gene Name: | PHD finger protein 2 |
Gene Aliases: |
CENP-35, GRC5, JHDM1E |
Location: |
Chr.9:93576627-93679587 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHF2 | NM_005392.3 | NP_005383.3 | ||
| XM_005252051.2 | XP_005252108.1 | |||
| XM_006717143.2 | XP_006717206.1 | |||
| AF043725.1 | AAD21791.1 | |||
| AL834263.1 | CAD38938.1 | |||
| BC064613.1 | AAH64613.1 | |||
| CB990295.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv824984 | Chr.9:93548651 - 93612740 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map