Assay Details
Target Gene Details
Entrez Gene ID: | 11092 |
Gene Name: | sperm acrosome associated 9 |
Gene Aliases: |
C9orf9, Mast |
Location: |
Chr.9:132878320-132890314 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPACA9 | NM_001316897.1 | NP_001303826.1 | ||
| NM_001316898.1 | NP_001303827.1 | |||
| NM_001316900.1 | NP_001303829.1 | |||
| NM_018956.4 | NP_061829.3 | |||
| NR_133631.1 | ||||
| XM_017014231.1 | XP_016869720.1 | |||
| AJ011375.1 | CAB66158.1 | |||
| AK075259.1 | BAC11503.1 | |||
| BC012940.1 | AAH12940.1 | |||
| CK906174.1 | ||||
| CK906175.1 | ||||
| CR457210.1 | CAG33491.1 | |||
| DA134573.1 | ||||
| DA281620.1 | ||||
| HY024188.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| nsv831743 | Chr.9:132820292 - 132982548 on Build GRCh38 | Loss |
|
| esv3621926 | Chr.9:132884570 - 132886716 on Build GRCh38 | Loss |
|
| esv3621927 | Chr.9:132885654 - 132894812 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map