Assay Details
Target Gene Details
Entrez Gene ID: | 286207 |
Gene Name: | cilia and flagella associated protein 157 |
Gene Aliases: |
C9orf117 |
Location: |
Chr.9:127706261-127716002 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CFAP157 | XM_006717064.3 | 1 | 618 | XP_006717127.2 |
| XM_011518559.2 | 1 | 621 | XP_011516861.2 | |
| XM_017014629.1 | 1 | 618 | XP_016870118.1 |
Target Gene Details
Entrez Gene ID: | 138428 |
Gene Name: | peptidyl-tRNA hydrolase 1 homolog |
Gene Aliases: |
C9orf115, PTH1 |
Location: |
Chr.9:127695024-127716714 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTRH1 | XM_017014277.1 | XP_016869766.1 |
More Information
Set Membership: |
|

Genomic Map