Assay Details
Target Gene Details
Entrez Gene ID: | 375748 |
Gene Name: | ERCC excision repair 6 like 2 |
Gene Aliases: |
BMFS2, C9orf102, RAD26L, SR278 |
Location: |
Chr.9:95875618-96041092 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERCC6L2 | NM_001010895.2 | NP_001010895.1 | ||
| NM_020207.4 | NP_064592.2 | |||
| XM_011518641.2 | XP_011516943.1 | |||
| XM_011518644.2 | XP_011516946.1 | |||
| XM_011518645.2 | XP_011516947.1 | |||
| XM_011518646.2 | XP_011516948.1 | |||
| XM_011518647.1 | XP_011516949.1 | |||
| XM_017014707.1 | XP_016870196.1 | |||
| AK307504.1 | ||||
| AK308409.1 | ||||
| AL389953.1 | CAB97543.1 | |||
| BC035183.1 | AAH35183.1 | |||
| BC140702.1 | ||||
| DB153432.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524894 | Chr.9:95875148 - 95877962 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map