Assay Details
Target Gene Details
Entrez Gene ID: | 347088 |
Gene Name: | adhesion G protein-coupled receptor D2 |
Gene Aliases: |
GPR144, PGR24 |
Location: |
Chr.9:124451087-124477795 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRD2 | XM_006717348.3 | XP_006717411.2 | ||
| AK022431.1 | 1 | 538 | ||
| AY278562.1 | AAP35064.1 | |||
| EF107718.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621722 | Chr.9:124378965 - 124497390 on Build GRCh38 | Gain |
|
| nsv615291 | Chr.9:124464413 - 124557925 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map