Assay Details
Target Gene Details
Entrez Gene ID: | 9328 |
Gene Name: | general transcription factor IIIC subunit 5 |
Gene Aliases: |
TFIIIC63, TFIIICepsilon, TFiiiC2-63 |
Location: |
Chr.9:133030675-133058503 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF3C5 | NM_001122823.1 | NP_001116295.1 | ||
| NM_001286709.1 | NP_001273638.1 | |||
| NM_012087.3 | NP_036219.2 | |||
| XM_005272234.3 | XP_005272291.1 | |||
| XM_011519201.1 | XP_011517503.1 | |||
| XM_017015315.1 | XP_016870804.1 | |||
| XM_017015316.1 | XP_016870805.1 | |||
| AF133124.1 | AAD41476.1 | |||
| AJ420443.1 | ||||
| AK000194.1 | BAA91001.1 | |||
| AK055092.1 | ||||
| AK097295.1 | BAC04993.1 | |||
| AK225761.1 | ||||
| AK293980.1 | ||||
| AK311481.1 | ||||
| AY007123.1 | AAG01991.1 | |||
| BC009741.2 | AAH09741.2 | |||
| BC011355.1 | AAH11355.1 | |||
| BC017337.2 | AAH17337.1 | |||
| BC030157.2 | AAH30157.2 | |||
| BQ673969.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| nsv466606 | Chr.9:133030314 - 133080285 on Build GRCh38 | Loss |
|
| nsv831744 | Chr.9:132980050 - 133174043 on Build GRCh38 | Gain+Loss |
|
| nsv508564 | Chr.9:133013830 - 133073076 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map