Assay Details
Target Gene Details
Entrez Gene ID: | 9630 |
Gene Name: | G protein subunit alpha 14 |
Gene Aliases: |
- |
Location: |
Chr.9:77423079-77648316 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNA14 | NM_004297.3 | NP_004288.1 | ||
| XM_011519224.1 | XP_011517526.1 | |||
| XM_017015341.1 | XP_016870830.1 | |||
| AF105201.1 | AAD17944.1 | |||
| AF493903.1 | AAM12617.1 | |||
| AK312460.1 | ||||
| BC027886.1 | AAH27886.1 |
Target Gene Details
Entrez Gene ID: | 101927422 |
Gene Name: | GNA14 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:77447649-77526762 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNA14-AS1 | NR_121184.1 | |||
| BI830360.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051079 | Chr.9:77361673 - 77471809 on Build GRCh38 | Gain |
|
| nsv1041647 | Chr.9:77211923 - 77537136 on Build GRCh38 | Gain |
|
| nsv614730 | Chr.9:77447358 - 77472115 on Build GRCh38 | Loss |
|
| nsv1036470 | Chr.9:77448138 - 77471048 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map