Assay Details
Target Gene Details
Entrez Gene ID: | 22927 |
Gene Name: | hyaluronan binding protein 4 |
Gene Aliases: |
IHABP-4, IHABP4, Ki-1/57, SERBP1L |
Location: |
Chr.9:96450155-96491336 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HABP4 | NM_014282.3 | NP_055097.2 | ||
| XM_005251812.3 | XP_005251869.1 | |||
| XM_017014484.1 | XP_016869973.1 | |||
| AF241831.1 | AAF62546.1 | |||
| AK025144.1 | ||||
| BC018788.1 | AAH18788.1 | |||
| BQ962055.1 | ||||
| U77327.1 | AAC31117.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv824998 | Chr.9:96394002 - 96467870 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs142851160] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map