Assay Details
Target Gene Details
Entrez Gene ID: | 360203 |
Gene Name: | glycosyltransferase 6 domain containing 1 |
Gene Aliases: |
GLTDC1, GT6M7 |
Location: |
Chr.9:135623656-135641216 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GLT6D1 | NM_182974.2 | NP_892019.2 | ||
| XM_011518633.1 | XP_011516935.1 | |||
| XM_011518634.1 | XP_011516936.1 | |||
| XM_011518635.1 | XP_011516937.1 | |||
| XM_011518636.1 | XP_011516938.1 | |||
| XM_011518637.1 | XP_011516939.1 | |||
| AY336054.1 | AAQ01588.1 |
More Information
Set Membership: |
|

Genomic Map