Assay Details
Target Gene Details
Entrez Gene ID: | 2776 |
Gene Name: | G protein subunit alpha q |
Gene Aliases: |
CMC1, G-ALPHA-q, GAQ, SWS |
Location: |
Chr.9:77716274-78031449 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GNAQ | NM_002072.4 | 7 | 6059 | NP_002063.2 |
| AK055619.1 | 1 | 1885 |
More Information
Set Membership: |
|

Genomic Map