Assay Details
Target Gene Details
Entrez Gene ID: | 27433 |
Gene Name: | torsin family 2 member A |
Gene Aliases: |
TORP1 |
Location: |
Chr.9:127731524-127735387 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TOR2A | NM_001085347.2 | NP_001078816.1 | ||
| NM_001134430.2 | NP_001127902.1 | |||
| NM_001134431.2 | NP_001127903.1 | |||
| NM_001252018.1 | NP_001238947.1 | |||
| NM_001252021.1 | NP_001238950.1 | |||
| NM_001252023.1 | NP_001238952.1 | |||
| NM_130459.3 | NP_569726.2 | |||
| XM_011518554.1 | XP_011516856.1 | |||
| AK027677.1 | BAB55288.1 | |||
| AK055034.1 | ||||
| AK075520.1 | BAC11667.1 | |||
| AK092388.1 | 1 | 1428 | ||
| AY358180.1 | AAQ88547.1 | |||
| BC062340.1 | AAH62340.1 | |||
| BC080527.1 | ||||
| BC100907.1 | ||||
| BC100908.2 | ||||
| BC100909.2 | ||||
| BC100910.1 | ||||
| BF115698.1 | ||||
| BM695589.1 | ||||
| BM772287.1 | ||||
| CN263138.1 | ||||
| DA430836.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv12879n54 | Chr.9:127731234 - 127824409 on Build GRCh38 | Loss |
|
| nsv615369 | Chr.9:127707924 - 127774438 on Build GRCh38 | Loss |
|
| nsv471323 | Chr.9:127727464 - 127845239 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map