Assay Details
Target Gene Details
Entrez Gene ID: | 28989 |
Gene Name: | N-terminal Xaa-Pro-Lys N-methyltransferase 1 |
Gene Aliases: |
AD-003, C9orf32, HOMT1A, METTL11A, NRMT, NTM1A |
Location: |
Chr.9:129608884-129636742 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NTMT1 | NM_001286796.1 | NP_001273725.1 | ||
| NM_001286797.1 | NP_001273726.1 | |||
| NM_001286798.1 | NP_001273727.1 | |||
| NM_001286799.1 | NP_001273728.1 | |||
| NM_001286800.1 | NP_001273729.1 | |||
| NM_001286801.1 | NP_001273730.1 | |||
| NM_001286802.1 | NP_001273731.1 | |||
| NM_001286803.1 | NP_001273732.1 | |||
| NM_014064.3 | NP_054783.2 | |||
| NR_104596.1 | ||||
| XM_005251939.3 | XP_005251996.1 | |||
| XM_017014642.1 | XP_016870131.1 | |||
| AF110776.1 | AAF14859.1 | |||
| AK290957.1 | ||||
| AK292332.1 | ||||
| AK298840.1 | ||||
| AK307056.1 | ||||
| AK313829.1 | ||||
| BC001396.2 | AAH01396.1 | |||
| BC033234.1 | AAH33234.1 | |||
| BE392289.1 | ||||
| BG468453.1 | ||||
| BI083998.1 | ||||
| BM009113.1 | ||||
| BM554138.1 | ||||
| BQ636193.1 | ||||
| BQ923165.1 | ||||
| CN406947.1 | ||||
| HY058177.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv469901 | Chr.9:129597897 - 129681820 on Build GRCh38 | Loss |
|
| nsv1046477 | Chr.9:129608081 - 129997996 on Build GRCh38 | Gain |
|
| nsv831731 | Chr.9:129514951 - 129634646 on Build GRCh38 | Gain+Loss |
|
| nsv951784 | Chr.9:129625222 - 129655421 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map