Assay Details
Target Gene Details
Entrez Gene ID: | 51104 |
Gene Name: | abhydrolase domain containing 17B |
Gene Aliases: |
C9orf77, CGI-67, FAM108B1 |
Location: |
Chr.9:71859461-71911245 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABHD17B | NM_001025780.2 | NP_001020951.1 | ||
| NM_016014.3 | NP_057098.2 | |||
| XM_006717134.3 | XP_006717197.2 | |||
| XM_006717135.1 | XP_006717198.1 | |||
| XM_017014787.1 | XP_016870276.1 | |||
| XM_017014788.1 | XP_016870277.1 | |||
| XM_017014789.1 | XP_016870278.1 | |||
| XM_017014790.1 | XP_016870279.1 | |||
| XM_017014791.1 | XP_016870280.1 | |||
| AF151825.1 | AAD34062.1 | |||
| AI564777.1 | ||||
| AK293060.1 | ||||
| BC038390.1 | ||||
| BC044576.1 | AAH44576.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761535 | Chr.9:71872257 - 71920586 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map