Assay Details
Target Gene Details
Entrez Gene ID: | 101929302 |
Gene Name: | RFX3 antisense RNA 1 |
Gene Aliases: |
CVAT7 |
Location: |
Chr.9:3668086-3691756 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFX3-AS1 | NR_121586.1 | 5 | 979 | |
| AW977824.1 | 1 | 131 | ||
| H56575.1 | 2 | 250 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3619316 | Chr.9:3688604 - 3695820 on Build GRCh38 | Loss |
|
| dgv7378n100 | Chr.9:3594189 - 3732530 on Build GRCh38 | Loss |
|

Genomic Map