Assay Details
Target Gene Details
Entrez Gene ID: | 57720 |
Gene Name: | G protein-coupled receptor 107 |
Gene Aliases: |
GCDRP, LUSTR1, bA138E2.2 |
Location: |
Chr.9:130053706-130140169 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR107 | NM_001136557.1 | NP_001130029.1 | ||
| NM_001136558.1 | NP_001130030.1 | |||
| NM_001287346.1 | NP_001274275.1 | |||
| NM_020960.4 | NP_066011.2 | |||
| XM_017014953.1 | XP_016870442.1 | |||
| XM_017014954.1 | XP_016870443.1 | |||
| AB046844.1 | BAB13450.1 | |||
| AF376725.1 | AAK57695.1 | |||
| AK026244.1 | BAB15408.1 | |||
| AK225456.1 | ||||
| AK293092.1 | ||||
| AL834359.1 | CAI46205.1 | |||
| BC110518.2 | AAI10519.1 | |||
| BC143655.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1046307 | Chr.9:130029993 - 130219186 on Build GRCh38 | Gain |
|
| nsv831732 | Chr.9:129920648 - 130103900 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs75397884] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map