Assay Details
Target Gene Details
Entrez Gene ID: | 100616278 |
Gene Name: | microRNA 4540 |
Gene Aliases: |
- |
Location: |
Chr.9:36864254-36864308 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR4540 | NR_039766.1 |
Target Gene Details
Entrez Gene ID: | 5079 |
Gene Name: | paired box 5 |
Gene Aliases: |
ALL3, BSAP |
Location: |
Chr.9:36833274-37035949 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6531 | Chr.9:36832805 - 36876608 on Build GRCh38 | Deletion |
|
| nsv466387 | Chr.9:36856271 - 36885586 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs141426259,rs576786508] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map