Assay Details
Target Gene Details
Entrez Gene ID: | 7010 |
Gene Name: | TEK receptor tyrosine kinase |
Gene Aliases: |
CD202B, TIE-2, TIE2, VMCM, VMCM1 |
Location: |
Chr.9:27109141-27230178 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TEK | NM_000459.4 | NP_000450.2 | ||
| NM_001290077.1 | NP_001277006.1 | |||
| NM_001290078.1 | NP_001277007.1 | |||
| XM_005251561.1 | XP_005251618.1 | |||
| XM_005251563.1 | XP_005251620.1 | |||
| AB086825.1 | BAC45250.1 | |||
| AB208796.1 | BAD92033.1 | |||
| AK291775.1 | ||||
| AK294887.1 | ||||
| AK295043.1 | ||||
| AK308447.1 | ||||
| BC035514.1 | ||||
| CA392256.1 | ||||
| DC322431.1 | ||||
| DC324016.1 | ||||
| EU826591.1 | ||||
| EU826592.1 | ||||
| L06139.1 | AAA61139.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7487n100 | Chr.9:27073652 - 27264578 on Build GRCh38 | Gain |
|
| nsv517269 | Chr.9:26945978 - 27137244 on Build GRCh38 | Gain |
|
| nsv1015259 | Chr.9:26347305 - 27643043 on Build GRCh38 | Gain |
|
| esv3891647 | Chr.9:26743024 - 27123262 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map