Assay Details
Target Gene Details
Entrez Gene ID: | 158067 |
Gene Name: | adenylate kinase 8 |
Gene Aliases: |
AK 8, C9orf98, DDX31 |
Location: |
Chr.9:132725574-132878811 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AK8 | NM_001317958.1 | NP_001304887.1 | ||
| NM_001317959.1 | NP_001304888.1 | |||
| NM_152572.2 | NP_689785.1 | |||
| XM_005272169.2 | XP_005272226.1 | |||
| XM_006716965.2 | XP_006717028.1 | |||
| XM_011518278.2 | XP_011516580.1 | |||
| XM_017014308.1 | XP_016869797.1 | |||
| AK057266.1 | BAB71402.1 | |||
| AK093446.1 | BAC04168.1 | |||
| AK292186.1 | ||||
| BC034776.1 | AAH34776.1 | |||
| BC050576.1 | AAH50576.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1051206 | Chr.9:132723216 - 132863839 on Build GRCh38 | Gain |
|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| dgv1513e214 | Chr.9:132713668 - 132865324 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map