Assay Details
Target Gene Details
Entrez Gene ID: | 1902 |
Gene Name: | lysophosphatidic acid receptor 1 |
Gene Aliases: |
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1 |
Location: |
Chr.9:110873252-111039259 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
More Information
Additional Information:
For this assay, SNP(s) [rs67686996] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map