Assay Details
Target Gene Details
Entrez Gene ID: | 23012 |
Gene Name: | serine/threonine kinase 38 like |
Gene Aliases: |
NDR2 |
Location: |
Chr.12:27244143-27325959 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK38L | NM_015000.3 | NP_055815.1 | ||
| XM_005253342.3 | XP_005253399.1 | |||
| XM_006719058.3 | XP_006719121.1 | |||
| XM_006719059.3 | XP_006719122.1 | |||
| XM_017019036.1 | XP_016874525.1 | |||
| AB023182.2 | ||||
| AK291055.1 | ||||
| AK304755.1 | ||||
| BC028603.1 | AAH28603.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1406n100 | Chr.12:27128236 - 27640822 on Build GRCh38 | Gain |
|
| dgv263e214 | Chr.12:27152627 - 27304231 on Build GRCh38 | Gain |
|
| dgv2417n54 | Chr.12:27068022 - 27638988 on Build GRCh38 | Gain |
|
| nsv436825 | Chr.12:27135971 - 27642579 on Build GRCh38 | Insertion |
|
| esv2745664 | Chr.12:27017516 - 27820117 on Build GRCh38 | Deletion |
|
| dgv271e199 | Chr.12:27089700 - 27631451 on Build GRCh38 | Deletion |
|
| nsv521872 | Chr.12:27114021 - 27635541 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs140667295] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map