Assay Details
Target Gene Details
Entrez Gene ID: | 283420 |
Gene Name: | C-type lectin domain family 9 member A |
Gene Aliases: |
CD370, DNGR-1, DNGR1, UNQ9341 |
Location: |
Chr.12:10030677-10066030 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLEC9A | NM_207345.3 | NP_997228.1 | ||
| AK307335.1 | ||||
| AY358265.1 | AAQ88632.1 | |||
| DB090462.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs76676564] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map