Assay Details
Target Gene Details
Entrez Gene ID: | 51768 |
Gene Name: | transmembrane 7 superfamily member 3 |
Gene Aliases: |
- |
Location: |
Chr.12:26966519-27014427 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TM7SF3 | XM_005253391.3 | 11 | 7199 | XP_005253448.1 |
| XM_011520690.2 | 11 | 7194 | XP_011518992.1 | |
| XM_017019463.1 | 12 | 7442 | XP_016874952.1 | |
| XM_017019464.1 | 10 | 7004 | XP_016874953.1 | |
| XM_017019465.1 | 9 | 6851 | XP_016874954.1 | |
| XM_017019466.1 | 10 | 7018 | XP_016874955.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892165 | Chr.12:26965680 - 26971722 on Build GRCh38 | Loss |
|
| esv3892164 | Chr.12:26945269 - 26971722 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map