Assay Details
Target Gene Details
Entrez Gene ID: | 121340 |
Gene Name: | Sp7 transcription factor |
Gene Aliases: |
OI11, OI12, OSX, osterix |
Location: |
Chr.12:53326575-53344793 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SP7 | NM_001300837.1 | NP_001287766.1 | ||
| XM_011537900.2 | XP_011536202.1 | |||
| HY020346.1 |
More Information
Set Membership: |
|

Genomic Map