Assay Details
Target Gene Details
Entrez Gene ID: | 11171 |
Gene Name: | serine/threonine kinase receptor associated protein |
Gene Aliases: |
MAWD, PT-WD, UNRIP |
Location: |
Chr.12:15882354-15903478 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STRAP | NM_007178.3 | NP_009109.3 | ||
| XM_005253294.1 | XP_005253351.1 | |||
| AB024327.1 | BAA75544.1 | |||
| AF161496.1 | AAF29111.1 | |||
| AJ010025.1 | CAB38041.1 | |||
| AK297942.1 | ||||
| AK312295.1 | ||||
| AK315891.1 | ||||
| AL136691.1 | CAB66626.1 | |||
| AY049776.1 | AAL15433.1 | |||
| BC000162.2 | AAH00162.1 | |||
| BC062306.1 | AAH62306.1 | |||
| BT020044.1 | AAV38847.1 | |||
| BT020045.1 | AAV38848.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760899 | Chr.12:15643377 - 15943563 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map