Assay Details
Target Gene Details
Entrez Gene ID: | 5204 |
Gene Name: | prefoldin subunit 5 |
Gene Aliases: |
MM-1, MM1, PFD5 |
Location: |
Chr.12:53295451-53299450 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PFDN5 | NM_002624.3 | NP_002615.2 | ||
| NM_145897.2 | NP_665904.1 | |||
| AB055803.1 | BAB32644.1 | |||
| AB055804.1 | BAB32645.1 | |||
| AB055805.1 | BAB32646.1 | |||
| AK024094.1 | 3 | 695 | ||
| AK292623.1 | ||||
| BC062671.1 | AAH62671.1 | |||
| BE392248.1 | ||||
| BT007195.1 | AAP35859.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv517066 | Chr.12:53006394 - 53340722 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs149989496] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map