Assay Details
Target Gene Details
Entrez Gene ID: | 23074 |
Gene Name: | UHRF1 binding protein 1 like |
Gene Aliases: |
SHIP164 |
Location: |
Chr.12:100026251-100142864 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UHRF1BP1L | XM_017019047.1 | 21 | 6380 | XP_016874536.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2826n54 | Chr.12:99884262 - 100152272 on Build GRCh38 | Loss |
|
| esv3892256 | Chr.12:100007917 - 100225312 on Build GRCh38 | Loss |
|
| nsv523049 | Chr.12:100017297 - 100039015 on Build GRCh38 | Loss |
|
| esv3630541 | Chr.12:99884244 - 100071236 on Build GRCh38 | Gain |
|
| nsv559964 | Chr.12:99872025 - 100538490 on Build GRCh38 | Gain |
|
| nsv559994 | Chr.12:100002546 - 100057456 on Build GRCh38 | Loss |
|

Genomic Map