Assay Details
Target Gene Details
Entrez Gene ID: | 55010 |
Gene Name: | PARP1 binding protein |
Gene Aliases: |
AROM, C12orf48, PARI |
Location: |
Chr.12:102120171-102197520 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
More Information
Set Membership: |
|

Genomic Map