Assay Details
Target Gene Details
Entrez Gene ID: | 11226 |
Gene Name: | polypeptide N-acetylgalactosaminyltransferase 6 |
Gene Aliases: |
GALNAC-T6, GalNAcT6 |
Location: |
Chr.12:51351870-51391955 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALNT6 | NM_007210.3 | NP_009141.2 | ||
| XM_005268607.1 | XP_005268664.1 | |||
| XM_006719214.2 | XP_006719277.1 | |||
| XM_011537819.2 | XP_011536121.1 | |||
| XM_011537822.2 | XP_011536124.1 | |||
| XM_017018744.1 | XP_016874233.1 | |||
| XM_017018745.1 | XP_016874234.1 | |||
| AB265820.1 | ||||
| AK074658.1 | BAC11118.1 | |||
| AK296852.1 | ||||
| BC035822.1 | AAH35822.2 | |||
| BC114505.1 | AAI14506.1 | |||
| Y08565.1 | CAA69876.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832408 | Chr.12:51302821 - 51477555 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map