Assay Details
Target Gene Details
Entrez Gene ID: | 121227 |
Gene Name: | leucine rich repeats and immunoglobulin like domains 3 |
Gene Aliases: |
LIG3 |
Location: |
Chr.12:58869351-58920540 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRIG3 | XM_011537893.2 | 19 | 4215 | XP_011536195.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759902 | Chr.12:58628947 - 58929264 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map