Assay Details
Target Gene Details
Entrez Gene ID: | 10728 |
Gene Name: | prostaglandin E synthase 3 |
Gene Aliases: |
P23, TEBP, cPGES |
Location: |
Chr.12:56663341-56689250 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGES3 | NM_001282601.1 | NP_001269530.1 | ||
| NM_001282602.1 | NP_001269531.1 | |||
| NM_001282603.1 | NP_001269532.1 | |||
| NM_001282604.1 | NP_001269533.1 | |||
| NM_001282605.1 | NP_001269534.1 | |||
| NM_006601.6 | NP_006592.3 | |||
| NR_104219.1 | 1 | 676 | ||
| XM_005268576.4 | 1 | 1929 | XP_005268633.1 | |
| XM_006719199.2 | 1 | 688 | XP_006719262.1 | |
| XM_011537773.2 | XP_011536075.1 | |||
| XM_011537774.2 | 1 | 688 | XP_011536076.1 | |
| XM_017018716.1 | 1 | 688 | XP_016874205.1 | |
| AK098214.1 | 1 | 676 | ||
| AK291945.1 | ||||
| AK293139.1 | ||||
| AK295208.1 | ||||
| AK298147.1 | ||||
| AK298160.1 | ||||
| AL603017.1 | ||||
| BC003005.1 | AAH03005.1 | |||
| BM541503.1 | ||||
| BP194632.1 | ||||
| BU167210.1 | ||||
| L24804.1 | AAA18537.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map