Assay Details
Target Gene Details
Entrez Gene ID: | 83892 |
Gene Name: | potassium channel tetramerization domain containing 10 |
Gene Aliases: |
BTBD28, MSTP028, ULRO61, hBACURD3 |
Location: |
Chr.12:109448655-109477544 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCTD10 | NM_001317395.1 | NP_001304324.1 | ||
| NM_001317399.1 | NP_001304328.1 | |||
| NM_031954.4 | NP_114160.1 | |||
| NR_133897.1 | ||||
| NR_133898.1 | ||||
| DA440672.1 |
Target Gene Details
Entrez Gene ID: | 89910 |
Gene Name: | ubiquitin protein ligase E3B |
Gene Aliases: |
BPIDS, KOS |
Location: |
Chr.12:109477410-109543628 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBE3B | XM_005253987.2 | 1 | 168 | XP_005254044.1 |
| XM_006719681.3 | 1 | 168 | XP_006719744.1 | |
| XM_006719682.2 | 1 | 168 | XP_006719745.1 | |
| XM_011538959.2 | 1 | 168 | XP_011537261.1 | |
| XM_011538961.1 | 1 | 168 | XP_011537263.1 | |
| XM_017020195.1 | 1 | 168 | XP_016875684.1 | |
| XM_017020196.1 | 1 | 168 | XP_016875685.1 |
More Information
Set Membership: |
|

Genomic Map