Assay Details
Target Gene Details
Entrez Gene ID: | 387893 | 
Gene Name: | lysine methyltransferase 5A | 
Gene Aliases: | 
	
		PR-Set7, SET07, SET8, SETD8 | 
Location: | 
		
			Chr.12:123383778-123409357 on Build GRCh38 | 
		Assay Gene Location: | Within Intron 7 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| KMT5A | NM_001324504.1 | NP_001311433.1 | ||
| NM_001324505.1 | NP_001311434.1 | |||
| NM_001324506.1 | NP_001311435.1 | |||
| NM_020382.4 | NP_065115.3 | |||
| NR_136910.1 | ||||
| XM_006719394.3 | XP_006719457.1 | |||
| XM_011538333.2 | XP_011536635.1 | |||
| AK292645.1 | ||||
| AY102937.1 | AAM47033.1 | |||
| BC050346.1 | AAH50346.1 | |||
| HQ259683.1 | 
More Information
Set Membership: | 
		   
					
						
							 | 
				

Genomic Map