Assay Details
Target Gene Details
Entrez Gene ID: | 4673 |
Gene Name: | nucleosome assembly protein 1 like 1 |
Gene Aliases: |
NAP1, NAP1L, NRP |
Location: |
Chr.12:76045383-76085033 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAP1L1 | NM_004537.5 | 15 | 4060 | NP_004528.1 |
| NM_139207.3 | 16 | 3386 | NP_631946.1 | |
| AK021751.1 | 1 | 226 | ||
| CD000987.1 | ||||
| DA137494.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv973093 | Chr.12:76045528 - 76048557 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map