Assay Details
Target Gene Details
Entrez Gene ID: | 10818 |
Gene Name: | fibroblast growth factor receptor substrate 2 |
Gene Aliases: |
FRS1A, FRS2A, FRS2alpha, SNT, SNT-1, SNT1 |
Location: |
Chr.12:69470349-69579793 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FRS2 | NM_001042555.2 | NP_001036020.1 | ||
| NM_001278351.1 | NP_001265280.1 | |||
| NM_001278353.1 | NP_001265282.1 | |||
| NM_001278354.1 | NP_001265283.1 | |||
| NM_001278355.1 | NP_001265284.1 | |||
| NM_001278356.1 | NP_001265285.1 | |||
| NM_001278357.1 | NP_001265286.1 | |||
| NM_006654.4 | NP_006645.3 | |||
| XM_017018717.1 | XP_016874206.1 | |||
| XM_017018718.1 | XP_016874207.1 | |||
| AK312477.1 | ||||
| BC021562.1 | AAH21562.1 | |||
| BP217337.1 | ||||
| BP230333.1 | ||||
| BP280775.1 | ||||
| BP311537.1 | ||||
| BP363404.1 | ||||
| BU166503.1 | ||||
| DA189114.1 | ||||
| DA311708.1 | ||||
| DA721439.1 | ||||
| DB081843.1 | ||||
| DR003634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559225 | Chr.12:69468445 - 69474630 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76177064] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map