Assay Details
Target Gene Details
Entrez Gene ID: | 56938 |
Gene Name: | aryl hydrocarbon receptor nuclear translocator like 2 |
Gene Aliases: |
BMAL2, CLIF, MOP9, PASD9, bHLHe6 |
Location: |
Chr.12:27332854-27425813 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARNTL2 | NM_001248002.1 | 16 | 6305 | NP_001234931.1 |
| NM_001248003.1 | 15 | 6236 | NP_001234932.1 | |
| NM_001248004.1 | 15 | 6203 | NP_001234933.1 | |
| NM_001248005.1 | 14 | 6127 | NP_001234934.1 | |
| NM_020183.4 | 17 | 6347 | NP_064568.3 | |
| XM_006719112.3 | 16 | 6306 | XP_006719175.1 | |
| XM_006719114.3 | 16 | 6037 | XP_006719177.1 | |
| XM_011520766.2 | 17 | 6348 | XP_011519068.1 | |
| XM_011520768.2 | 16 | 6256 | XP_011519070.1 | |
| XM_011520769.2 | 16 | 6246 | XP_011519071.1 | |
| XM_011520770.2 | 17 | 6166 | XP_011519072.1 | |
| XM_011520771.2 | 17 | 6132 | XP_011519073.1 | |
| XM_017019666.1 | 14 | 6077 | XP_016875155.1 | |
| XM_017019667.1 | 17 | 6364 | XP_016875156.1 | |
| XM_017019668.1 | 17 | 6161 | XP_016875157.1 | |
| XM_017019669.1 | 16 | 6008 | XP_016875158.1 |
Target Gene Details
Entrez Gene ID: | 101928646 |
Gene Name: | ARNTL2 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:27389789-27446634 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARNTL2-AS1 | NR_109975.1 | |||
| BU568793.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv1406n100 | Chr.12:27128236 - 27640822 on Build GRCh38 | Gain |
|
| dgv2417n54 | Chr.12:27068022 - 27638988 on Build GRCh38 | Gain |
|
| nsv650 | Chr.12:27410480 - 27444459 on Build GRCh38 | Insertion |
|
| nsv436825 | Chr.12:27135971 - 27642579 on Build GRCh38 | Insertion |
|
| esv2745664 | Chr.12:27017516 - 27820117 on Build GRCh38 | Deletion |
|
| nsv832357 | Chr.12:27399918 - 27439033 on Build GRCh38 | Gain |
|
| dgv271e199 | Chr.12:27089700 - 27631451 on Build GRCh38 | Deletion |
|
| nsv521872 | Chr.12:27114021 - 27635541 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map