Assay Details
Target Gene Details
Entrez Gene ID: | 140707 |
Gene Name: | BRI3 binding protein |
Gene Aliases: |
BNAS1, HCCR-1, HCCR-2, HCCRBP-1, KG19 |
Location: |
Chr.12:124993648-125051845 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BRI3BP | XM_011537940.2 | XP_011536242.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1047470 | Chr.12:125009120 - 125044073 on Build GRCh38 | Loss |
|
| nsv1051020 | Chr.12:125015140 - 125809096 on Build GRCh38 | Gain |
|
| esv3892269 | Chr.12:125024464 - 125055560 on Build GRCh38 | Gain |
|

Genomic Map