Assay Details
Target Gene Details
Entrez Gene ID: | 225 |
Gene Name: | ATP binding cassette subfamily D member 2 |
Gene Aliases: |
ABC39, ALDL1, ALDR, ALDRP, hALDR |
Location: |
Chr.12:39530966-39651333 on Build GRCh38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ABCD2 | NM_005164.3 | NP_005155.1 | ||
| XM_011538027.2 | XP_011536329.1 | |||
| XM_017018992.1 | XP_016874481.1 | |||
| AJ000327.1 | CAA03994.1 | |||
| AK314254.1 | ||||
| BC104901.1 | AAI04902.1 | |||
| BC104903.1 | AAI04904.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1039645 | Chr.12:39509321 - 39628227 on Build GRCh38 | Loss |
|
| dgv2575n54 | Chr.12:39552552 - 39648418 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map